Learn More
6-Chloro-9-methyl-7-deazapurine, TRC
Product Code. 30366046
Change view
Click to view available options
Kvantitet:
2.5 g
Unit Size:
2.5g
Product Code. Kvantitet unitSize
30366046 2.5 g 2.5g
1 options
Product Code. 30366046 Supplier Toronto Research Chemicals Supplier No. TRCC3669952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 6-Chloro-9-methyl-7-deazapurine
CAS 7781-10-4
Molekylformel C7H6ClN3
Kvantitet 2.5 g
Synonym 4-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine, NSC 102340
InChI formel InChI=1S/C7H6ClN3/c1-11-3-2-5-6(8)9-4-10-7(5)11/h2-4H,1H3
LEDER Cn1ccc2c(Cl)ncnc12
IUPAC-namn 4-chloro-7-methylpyrrolo[2,3-d]pyrimidine
Molekylvikt (g/mol) 167.6
Formel vikt 167.025