Läs mer
6-Chlorohexanenitrile, TRC
Produktkod. 30364814
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364814 1 g
30363502 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364814 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3503301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6628-78-0
Molekylformel C6H10ClN
Molekylvikt (g/mol) 131.6
Synonym 6-Chlorohexanenitrile, 5-Chloropentyl Cyanide, 6-Chlorocapronitrile, NSC 59713
IUPAC-namn 6-chlorohexanenitrile
LEDER ClCCCCCC#N
Specifikationer
Kemiskt namn eller material 6-Chlorohexanenitrile
Kvantitet 1 g
InChI formel InChI=1S/C6H10ClN/c7-5-3-1-2-4-6-8/h1-5H2