Läs mer
6-Chloroisatin, TRC
Produktkod. 30352169
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352169 100 mg
30364004 50 mg
30349275 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC428808100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6341-92-0
Molekylformel C8H4NO2Cl
Molekylvikt (g/mol) 181.58
Synonym 6-Chloro-1H-indole-2,3-dione, 6-Chloroindole-2,3-dione, 6-Chloroindoline-2,3-dione, 6-Chloroisatin, NSC 48609
IUPAC-namn 6-chloro-1H-indole-2,3-dione
LEDER Clc1ccc2C(=O)C(=O)Nc2c1
Specifikationer
Kemiskt namn eller material 6-Chloroisatin
Kvantitet 100 mg
InChI formel InChI=1S/C8H4ClNO2/c9-4-1-2-5-6(3-4)10-8(12)7(5)11/h1-3H,(H,10,11,12)
Formel vikt 180.9931