missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7047561
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
4684-94-0
C6H4ClNO2
157.553
MFCD00155390
ZLKMOIHCHCMSFW-UHFFFAOYSA-N
6-chloropicolinic acid, 2-chloro-6-pyridinecarboxylic acid, 6-chloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-chloro, 2-chloropyridine-6-carboxylic acid, 6-chloro-2-picolinic acid, 6-cpa, picolinic acid, 6-chloro, 6-chloro-2-carboxy-pyridine, caswell no. 214b
20812
6-chloropyridine-2-carboxylic acid
C1=CC(=NC(=C1)Cl)C(=O)O
This item is not returnable. View return policy
4684-94-0
C6H4ClNO2
157.553
MFCD00155390
ZLKMOIHCHCMSFW-UHFFFAOYSA-N
6-chloropicolinic acid, 2-chloro-6-pyridinecarboxylic acid, 6-chloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-chloro, 2-chloropyridine-6-carboxylic acid, 6-chloro-2-picolinic acid, 6-cpa, picolinic acid, 6-chloro, 6-chloro-2-carboxy-pyridine, caswell no. 214b
20812
6-chloropyridine-2-carboxylic acid
C1=CC(=NC(=C1)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 194°C
Quantity 250 mg
Beilstein 115849
Formula Weight 157.56
Percent Purity 98%
Chemical Name or Material 6-Chloropyridine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'