Läs mer
6-Deoxypenciclovir hydrochloride, TRC
Produktkod. 30134167
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134167 500 mg
30137647 100 mg
30010932 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134167 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD247645500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 246021-75-0
Molekylformel C10 H15 N5 O2 . Cl H
Molekylvikt (g/mol) 273.72
Synonym 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, hydrochloride (1:1), 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, monohydrochloride (9CI)
IUPAC-namn 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol;hydrochloride
LEDER Cl.Nc1ncc2ncn(CCC(CO)CO)c2n1
Specifikationer
Kemiskt namn eller material 2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C10H15N5O2.ClH/c11-10-12-3-8-9(14-10)15(6-13-8)2-1-7(4-16)5-17;/h3,6-7,16-17H,1-2,4-5H2,(H2,11,12,14);1H
Formel vikt 273.0993