Läs mer
6-Des(diethanolamino)-6-chloro Dipyridamole, TRC
Produktkod. 30352988
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352988 100 mg
30358073 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288935100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54093-92-4
Molekylformel C20 H30 Cl N7 O2
Molekylvikt (g/mol) 435.95
Synonym 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Imp. C (EP), Dipyridamole USP Related Compound C, Dipyridamole USP RC C, 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Related Compound C, Dipyridamole Impurity C, 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Imp. C (EP)
IUPAC-namn 2-[[2-chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
LEDER OCCN(CCO)c1nc(N2CCCCC2)c3nc(Cl)nc(N4CCCCC4)c3n1
Specifikationer
Kemiskt namn eller material 2,2'-((6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol
Kvantitet 100 mg
InChI formel InChI=1S/C20H30ClN7O2/c21-19-22-15-16(17(24-19)26-7-3-1-4-8-26)23-20(28(11-13-29)12-14-30)25-18(15)27-9-5-2-6-10-27/h29-30H,1-14H2
Formel vikt 435.215