Läs mer
6-Desacetyl-N-Boc Palbociclib, TRC
Produktkod. 30366881
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366881 1 mg
30351616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2887131MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 571189-65-6
Molekylformel C27H35N7O3
Molekylvikt (g/mol) 505.61
Synonym 4-[6-[(8-Cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino]pyridin-3-yl]piperazine-1-carboxylic Acid tert-Butyl Ester
LEDER CC1=CC(=O)N(C2CCCC2)c3nc(Nc4ccc(cn4)N5CCN(CC5)C(=O)OC(C)(C)C)ncc13
Specifikationer
Kemiskt namn eller material 6-Desacetyl-N-Boc Palbociclib
Kvantitet 1 mg
InChI formel InChI=1S/C27H35N7O3/c1-18-15-23(35)34(19-7-5-6-8-19)24-21(18)17-29-25(31-24)30-22-10-9-20(16-28-22)32-11-13-33(14-12-32)26(36)37-27(2,3)4/h9-10,15-17,19H,5-8,11-14H2,1-4H3,(H,28,29,30,31)