Läs mer
6-Desacetyl Palbociclib, TRC
Produktkod. 30133141
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133141 50 mg
30133348 5 mg
30012703 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133141 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28871850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 571190-22-2
Molekylformel C22H27N7O
Molekylvikt (g/mol) 405.5
Synonym 8-Cyclopentyl-5-methyl-2-[[(5-piperazin-1-ylpyridin-2-yl)]amino]-8H-pyrido[2,3-d]pyrimidin-7-one;
LEDER CC1=CC(=O)N(C2CCCC2)c3nc(Nc4ccc(cn4)N5CCNCC5)ncc13
Specifikationer
Kemiskt namn eller material 6-Desacetyl Palbociclib
Analyt- eller komponentnamn 6-Desacetyl Palbociclib
Frakt skick Room Temperature
Lösningstyp Neat
Industry Type Pharmaceutical