Läs mer
6-Deschloro Cyproterone Acetate, TRC
Produktkod. 30365229
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30365229 50 mg
1 options
Produktkod. 30365229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28852050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17-Hydroxy-1Alpha,2Alpha-methylenepregna-4,6-diene-3,20-dione 17-Acetate
CAS 2701-50-0
Molekylformel C24 H30 O4
Kvantitet 50 mg
Synonym 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 17-(acetyloxy)-1,2-dihydro-, (1β,2β)-, 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 1β,2β-dihydro-17-hydroxy-, acetate (8CI), Pregna-4,6-diene-3,20-dione, 17-hydroxy-1α,2α-methylene-, acetate (6CI,7
InChI formel InChI=1S/C24H30O4/c1-13(25)24(28-14(2)26)10-8-18-16-6-5-15-11-21(27)17-12-20(17)23(15,4)19(16)7-9-22(18,24)3/h5-6,11,16-20H,7-10,12H2,1-4H3/t16-,17+,18-,19-,20-,22-,23-,24-/m0/s1
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=CC4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Molekylvikt (g/mol) 382.493
Formel vikt 382.214