Läs mer
6’-Desmethoxy-6’-hydroxy Noscapine, TRC
Produktkod. 30365043
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365043 100 mg
30363267 10 mg
30362422 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD235690100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 220875-41-2
Molekylformel C21H21NO7
Molekylvikt (g/mol) 399.4
Synonym (3S)-6-Hydroxy-7-methoxy-3-[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-1(3H)-isobenzofuranone, Noscapine Impurity
IUPAC-namn (S)-6-hydroxy-7-methoxy-3-((R)-4-methoxy-6-methyl-5,6,7,8-tetrahydro-[1,3]dioxolo[4,5-g]isoquinolin-5-yl)isobenzofuran-1(3H)-one
LEDER [H][C@@]1(C2=C(C3=C(C=C2CCN1C)OCO3)OC)[C@]4([H])C5=CC=C(C(OC)=C5C(O4)=O)O
Specifikationer
Kemiskt namn eller material Noscapine Impurity 11
Kvantitet 100 mg
InChI formel InChI=1S/C21H21NO7/c1-22-7-6-10-8-13-19(28-9-27-13)20(26-3)14(10)16(22)17-11-4-5-12(23)18(25-2)15(11)21(24)29-17/h4-5,8,16-17,23H,6-7,9H2,1-3H3/t16-,17+/m1/s1
Formel vikt 399.13