Läs mer
6-Despiperidinyl-6-(alogliptin-Namino-yl) Alogliptin, TRC
Produktkod. 30363301
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363301 100 mg
30354761 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363301 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD297155100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1268836-55-0
Molekylformel C31H30N8O4
Molekylvikt (g/mol) 578.62
Synonym 2-[[6-[(3R)-3-[[3-[(2-cyanophenyl)methyl]-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-4-pyrimidinyl]amino]-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-benzonitrile
IUPAC-namn (R)-2-((6-(3-((3-(2-cyanobenzyl)-1-methyl-2,6-dioxo-1,2,3,6-tetrahydropyrimidin-4-yl)amino)piperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile
LEDER C(N1C(=CC(=O)N(C)C1=O)N2C[C@H](NC=3N(CC4=C(C#N)C=CC=C4)C(=O)N(C)C(=O)C3)CCC2)C5=C(C#N)C=CC=C5
Specifikationer
Kemiskt namn eller material 6-Despiperidinyl-6-(alogliptin-Namino-yl) Alogliptin
Kvantitet 100 mg
InChI formel InChI=1S/C31H30N8O4/c1-35-28(40)14-26(38(30(35)42)18-23-10-5-3-8-21(23)16-32)34-25-12-7-13-37(20-25)27-15-29(41)36(2)31(43)39(27)19-24-11-6-4-9-22(24)17-33/h3-6,8-11,14-15,25,34H,7,12-13,18-20H2,1-2H3/t25-/m1/s1
Formel vikt 578.24