missing translation for 'onlineSavingsMsg'
Learn More

6-(Diethylamino)-3-pyridinylaldehyde, 97%, Thermo Scientific™

Catalog Number p-3075816
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
578726-67-7
C10H14N2O
178.24
MFCD06200894
PCISUIMHLSRKGR-UHFFFAOYSA-N
6-diethylamino pyridine-3-carbaldehyde, 6-diethylamino-3-pyridinylaldehyde, 6-diethylamino nicotinaldehyde, 6-diethylamino-3-pyridinecarboxaldehyde
2794806
6-(diethylamino)pyridine-3-carbaldehyde
CCN(CC)C1=NC=C(C=O)C=C1
This item is not returnable. View return policy
578726-67-7
C10H14N2O
178.24
MFCD06200894
PCISUIMHLSRKGR-UHFFFAOYSA-N
6-diethylamino pyridine-3-carbaldehyde, 6-diethylamino-3-pyridinylaldehyde, 6-diethylamino nicotinaldehyde, 6-diethylamino-3-pyridinecarboxaldehyde
2794806
6-(diethylamino)pyridine-3-carbaldehyde
CCN(CC)C1=NC=C(C=O)C=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 6-(Diethylamino)-3-pyridinylaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'