Läs mer
6-epi-Medroxy Progesterone 17-Acetate, TRC
Produktkod. 30134742
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134742 1 mg
30006636 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2035701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2242-65-1
Molekylformel C24 H34 O4
Molekylvikt (g/mol) 386.52
Synonym Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6β)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6β-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6β-methyl-, acetate (6CI), (6β)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 6β-Methyl-17α-hydroxyprogesterone acetate, NSC 119085
IUPAC-namn [(6R,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
LEDER C[C@@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)CCC(=O)C=C14
Specifikationer
Kemiskt namn eller material 6-Epimedroxyprogesterone acetate
Kvantitet 1 mg
InChI formel InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18-,19+,20+,22-,23+,24+/m1/s1
Formel vikt 386.2457