Läs mer
6-ethyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, TRC
Produktkod. 30332606
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30332606 10 mg
30343168 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30332606 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB44050510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-ethyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid
CAS 54881-17-3
Molekylformel C8H9NO3
Kvantitet 10 mg
InChI formel InChI=1S/C8H9NO3/c1-2-6-3-5(8(11)12)4-7(10)9-6/h3-4H,2H2,1H3,(H,9,10)(H,11,12)
LEDER CCC1=CC(=CC(=O)N1)C(=O)O
IUPAC-namn 2-ethyl-6-oxo-1H-pyridine-4-carboxylic acid
Molekylvikt (g/mol) 167.16
Formel vikt 167.0582