Läs mer
6-Ethyl-4-[4-(1,3,4-thiadiazol-2-yl)-1-piperazinyl]thieno[2,3-d]pyrimidine Hydrochloride, TRC
Produktkod. 30378188
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30378188 50 mg
1 options
Produktkod. 30378188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92674550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Ethyl-4-[4-(1,3,4-thiadiazol-2-yl)-1-piperazinyl]thieno[2,3-d]pyrimidine Hydrochloride
CAS 1359873-45-2
Molekylformel C14H17ClN6S2
Kvantitet 50 mg
Synonym 4-(4-(1,3,4-Thiadiazol-2-yl)piperazin-1-yl)-6-ethylthieno[2,3-d]pyrimidine Hydrochloride, MI-nc (hydrochloride)
InChI formel InChI=1S/C14H16N6S2.ClH/c1-2-10-7-11-12(15-8-16-13(11)22-10)19-3-5-20(6-4-19)14-18-17-9-21-14;/h7-9H,2-6H2,1H3;1H
LEDER Cl.CCc1cc2c(ncnc2s1)N3CCN(CC3)c4nncs4
Molekylvikt (g/mol) 368.91