Läs mer
MedChemExpress 6-FITC
Produktkod. 16477916
Encompass_Preferred
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16477916 100 mg
16431946 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16477916 Leverantör MedChemExpress Leverantörsnummer HY66020
MedChemExpress 6-FITC (6-Fluorescein Isothiocyanate) is a derivative of fluorescein used in wide-ranging applications including flow cytometry.
Specifikationer
Kvantitet 100 mg
Produkttyp Dye Reagents
Molekylvikt (g/mol) 389.38 g/mol
Löslighet DMSO : ≥ 35 mg/mL (89.89 mM)
Innehåll och lagring -20°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Frakt skick Room temperature
Mål HBV
CAS 3326-31-6
Synonym 6-Fluorescein Isothiocyanate