Läs mer
6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole, Mikromol™
Produktkod. 30051512
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051512 100 mg
30052591 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051512 Leverantör Mikromol Leverantörsnummer MM0491.36
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
CAS 2985481-41-0
Analyt- eller komponentnamn 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
Frakt skick Room Temperature
Molekylformel C12 H13 F N2 O2
Certifikat och efterlevnad ISO 17025
Synonym 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
InChI formel InChI=1S/C12H13FN2O2/c13-9-1-2-10-11(7-9)17-14-12(10)8-3-5-15(16)6-4-8/h1-2,7-8,16H,3-6H2
LEDER ON1CCC(CC1)c2noc3cc(F)ccc23
IUPAC-namn 6-fluoro-3-(1-hydroxypiperidin-4-yl)-1,2-benzoxazole
Molekylvikt (g/mol) 236.24
Kvantitet 100 mg
Formel vikt 236.0961 g/mol