missing translation for 'onlineSavingsMsg'
Learn More

6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7054747
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
88574-06-5
C21H23NO4
353.418
MFCD00077045
FPCPONSZWYDXRD-UHFFFAOYSA-N
fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
2756087
6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O
This item is not returnable. View return policy
88574-06-5
C21H23NO4
353.418
MFCD00077045
FPCPONSZWYDXRD-UHFFFAOYSA-N
fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
2756087
6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O

6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Linear Formula FmocNH(CH2)5CO2H
Quantity 1 g
Beilstein 5883220
Solubility Information Soluble in 1% acetic acid,water and 1mmol in 2mL DMF clearly soluble.
Formula Weight 353.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-(Fmoc-amino)hexanoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'