Learn More
6-Formyl-2-(methylsulfanyl)nicotinonitrile, 97%, Thermo Scientific™
Catalog Number p-4510530
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
175277-27-7
C8H6N2OS
178.21
MFCD00173824
UBGVOMZGBKLUOU-UHFFFAOYSA-N
6-formyl-2-methylsulfanyl nicotinonitrile, 3-cyano-2-methylthio pyridine-6-carboxaldehyde, 6-formyl-2-methylsulfanyl pyridine-3-carbonitrile, 6-formyl-2-methylthio nicotinonitrile, 6-formyl-2-methylthio pyridine-3-carbonitrile, 6-formyl-2-methylthio-3-pyridinecarbonitrile, 3-pyridinecarbonitrile,6-formyl-2-methylthio, 6-formyl-2-methylsulphanyl nicotinonitrile, 6-formyl-2-methylthiopyridine-3-carbonitrile, 6-formyl-2-methylsulfanyl-pyridine-3-carbonitrile
2799504
6-formyl-2-methylsulfanylpyridine-3-carbonitrile
CSC1=C(C=CC(C=O)=N1)C#N
175277-27-7
C8H6N2OS
178.21
MFCD00173824
UBGVOMZGBKLUOU-UHFFFAOYSA-N
6-formyl-2-methylsulfanyl nicotinonitrile, 3-cyano-2-methylthio pyridine-6-carboxaldehyde, 6-formyl-2-methylsulfanyl pyridine-3-carbonitrile, 6-formyl-2-methylthio nicotinonitrile, 6-formyl-2-methylthio pyridine-3-carbonitrile, 6-formyl-2-methylthio-3-pyridinecarbonitrile, 3-pyridinecarbonitrile,6-formyl-2-methylthio, 6-formyl-2-methylsulphanyl nicotinonitrile, 6-formyl-2-methylthiopyridine-3-carbonitrile, 6-formyl-2-methylsulfanyl-pyridine-3-carbonitrile
2799504
6-formyl-2-methylsulfanylpyridine-3-carbonitrile
CSC1=C(C=CC(C=O)=N1)C#N
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 6-Formyl-2-(methylsulfanyl)nicotinonitrile