Läs mer
(+)-[6]-Gingerol, TRC
Produktkod. 30137346
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137346 100 mg
18073220 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG387205100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23513-14-6
Molekylformel C17 H26 O4
Molekylvikt (g/mol) 294.39
Synonym 3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (S)-, 3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (S)-(+)- (8CI), (5S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, (+)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, (+)-[6]-Gingerol, (5S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)decan-3-one, (5S)-[6]-Gingerol, (S)-(+)-[6]Gingerol, (S)-[6]Gingerol, 3-Decanone 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5S)-, [6]-Gingerol, Gingerol
IUPAC-namn (5S)-5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)decan-3-one
LEDER CCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1
Specifikationer
Kemiskt namn eller material 6-Gingerol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C17H26O4/c1-3-4-5-6-14(18)12-15(19)9-7-13-8-10-16(20)17(11-13)21-2/h8,10-11,14,18,20H,3-7,9,12H2,1-2H3/t14-/m0/s1
Formel vikt 294.1831
Procent renhet >95%