Läs mer
6-Hexadecanoylamino-4-methylumbelliferyl Phosphorylcholine, TRC
Produktkod. 30004222
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004222 5 mg
30005022 10 mg
30004056 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2935905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 904315-61-3
Molekylformel C31 H51 N2 O7 P
Molekylvikt (g/mol) 594.72
Synonym Ethanaminium, 2-[[hydroxy[[4-methyl-2-oxo-6-[(1-oxohexadecyl)amino]-2H-1-benzopyran-7-yl]oxy]phosphinyl]oxy]-N,N,N-trimethyl-, inner salt, 2-[[Hydroxy[[4-methyl-2-oxo-6-[(1-oxohexadecyl)amino]-2H-1-benzopyran-7-yl]oxy]phosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, 6-Hexadecanoylamino-4-methylumbelliferyl phosphorylcholine
IUPAC-namn [6-(hexadecanoylamino)-4-methyl-2-oxochromen-7-yl] 2-(trimethylazaniumyl)ethyl phosphate
LEDER CCCCCCCCCCCCCCCC(=O)Nc1cc2C(=CC(=O)Oc2cc1OP(=O)([O-])OCC[N+](C)(C)C)C
Specifikationer
Kemiskt namn eller material 6-Hexadecanoylamino-4-methylumbelliferyl Phosphorylcholine
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C31H51N2O7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-30(34)32-27-23-26-25(2)22-31(35)39-28(26)24-29(27)40-41(36,37)38-21-20-33(3,4)5/h22-24H,6-21H2,1-5H3,(H-,32,34,36,37)
Formel vikt 594.3434
Procent renhet >95%