Läs mer
6-Hydroxy 2,4,5-Triaminopyrimidine Sulfate, TRC
Produktkod. 30403645
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30403645 250 mg -
30399352 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30403645 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH971210250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1603-02-7
Molekylformel C4 H7 N5 O4 S
Molekylvikt (g/mol) 221.19
Synonym 4-Pyrimidinol, 2,5,6-triamino-, hydrogen sulfate (7CI), 4-Pyrimidinol, 2,5,6-triamino-, hydrogen sulfate (ester) (8CI,9CI)
IUPAC-namn (2,5,6-triaminopyrimidin-4-yl) hydrogen sulfate
LEDER Nc1nc(N)c(N)c(OS(=O)(=O)O)n1
Specifikationer
Kemiskt namn eller material 6-Hydroxy 2,4,5-Triaminopyrimidine sulfate
Kvantitet 250 mg
InChI formel InChI=1S/C4H7N5O4S/c5-1-2(6)8-4(7)9-3(1)13-14(10,11)12/h5H2,(H,10,11,12)(H4,6,7,8,9)
Formel vikt 221.0219