Läs mer
6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, TRC
Produktkod. 30404106
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30404106 5 g
1 options
Produktkod. 30404106 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9507355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt
CAS 135-76-2
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H7 O4 S . Na
Kvantitet 5 g
Synonym 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt (1:1), 2-Naphthalenesulfonic acid, 6-hydroxy-, monosodium salt (8CI,9CI), 2-Hydroxy-6-naphthalenesulfonic acid sodium salt, 2-Naphthol-6-sodium sulfonate, 2-Naphthol-6-sulfonic acid sodium salt, 6-Sulfo-2-naphthol sodium salt, Schaeffer salt, Schaeffer's salt, Sodium 2-naphthol-6-sulfonate, Sodium 6-hydroxy-2-naphthalenesulfonate, Sodium β-naphthol-6-sulfonate
InChI formel InChI=1S/C10H8O4S.Na/c11-9-3-1-8-6-10(15(12,13)14)4-2-7(8)5-9;/h1-6,11H,(H,12,13,14);/q;+1/p-1
LEDER [Na+].Oc1ccc2cc(ccc2c1)S(=O)(=O)[O-]
IUPAC-namn sodium;6-hydroxynaphthalene-2-sulfonate
Molekylvikt (g/mol) 246.22
Formel vikt 245.9963