Läs mer
6-Hydroxy-3,4-dihydro-1H-quinoline-2-one, TRC
Produktkod. 30407758
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407758 1 g
30405588 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30407758 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9414231G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54197-66-9
Molekylformel C9 H9 N O2
Molekylvikt (g/mol) 163.17
Synonym 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 2(1H)-Quinolinone, 3,4-dihydro-6-hydroxy-Carbostyril, 3,4-dihydro-6-hydroxy- (7CI), Hydrocarbostyril, 6-hydroxy- (6CI), 2-Oxo-1,2,3,4-tetrahydroquinolin-6-ol, 3,4-Dihydro-6-hydroxy-2(1H)-quinolinone, 3,4-Dihydro-6-hydroxycarbostyril, 6-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline, 6-Hydroxy-3,4-dihydro-1H-quinolin-2-one, 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 6-Hydroxy-3,4-dihydrocarbostyril
IUPAC-namn 6-hydroxy-3,4-dihydro-1H-quinolin-2-one
LEDER Oc1ccc2NC(=O)CCc2c1
Specifikationer
Kemiskt namn eller material 6-Hydroxy-3,4-dihydro-1H-quinoline-2-one
Kvantitet 1 g
InChI formel InChI=1S/C9H9NO2/c11-7-2-3-8-6(5-7)1-4-9(12)10-8/h2-3,5,11H,1,4H2,(H,10,12)
Formel vikt 163.0633