Läs mer
6-Hydroxy-7-methoxycoumarin, TRC
Produktkod. 30404283
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30404283 25 mg
30392610 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30404283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH51734525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isoscopoletin
CAS 776-86-3
Molekylformel C10 H8 O4
Kvantitet 25 mg
Synonym 7-Methoxyesculetin, 6-Hydroxy-7-methoxycoumarin
InChI formel InChI=1S/C10H8O4/c1-13-9-5-8-6(4-7(9)11)2-3-10(12)14-8/h2-5,11H,1H3
LEDER COc1cc2OC(=O)C=Cc2cc1O
IUPAC-namn 6-hydroxy-7-methoxy-chromen-2-one
Molekylvikt (g/mol) 192.17
Formel vikt 192.0423