Läs mer
6-Hydroxy Chlorzoxazone-13C6, TRC
Produktkod. 30012870
Change view
Kvantitet:
0.25 mg
2.5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012870 2.5 mg 2.5mg
30010431 0.25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012870 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8251222.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 475295-90-0
Molekylformel C13C6H4ClNO3
Molekylvikt (g/mol) 191.52
Synonym 5-Chloro-6-hydroxy-2(3H)-benzoxazolone-13C6;
IUPAC-namn 5-Chloro-6-hydroxybenzo[d]oxazol-2(3H)-one-3a,4,5,6,7,7a-13C6
LEDER O[13C]1=[13CH][13C]2=[13C](NC(O2)=O)[13CH]=[13C]1Cl
Specifikationer
Kemiskt namn eller material 6-Hydroxy Chlorzoxazone-13C6
Kvantitet 2.5 mg
InChI formel InChI=1S/C7H4ClNO3/c8-3-1-4-6(2-5(3)10)12-7(11)9-4/h1-2,10H,(H,9,11)/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 191.0081