Läs mer
6-Hydroxy Chlorzoxazone, TRC
Produktkod. 30003756
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003756 5 mg
30004390 50 mg
30005634 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8251205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1750-45-4
Molekylformel C7 H4 Cl N O3
Molekylvikt (g/mol) 185.56
Synonym 2(3H)-Benzoxazolone, 5-chloro-6-hydroxy-, 2,6-Benzoxazolediol, 5-chloro- (6CI), 2-Benzoxazolinone, 5-chloro-6-hydroxy- (7CI,8CI), 6-Hydroxychloroxazone, 6-Hydroxychlorzoxazone, 6-Hydroxylchlorzoxazone, NSC 24955
IUPAC-namn 5-chloro-6-hydroxy-3H-1,3-benzoxazol-2-one
LEDER Oc1cc2OC(=O)Nc2cc1Cl
Specifikationer
Kemiskt namn eller material 6-Hydroxy Chlorzoxazone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C7H4ClNO3/c8-3-1-4-6(2-5(3)10)12-7(11)9-4/h1-2,10H,(H,9,11)
Formel vikt 184.988
Procent renhet >95%