missing translation for 'onlineSavingsMsg'
Learn More

6-(Hydroxymethyl)benzothiazole, 97%, Thermo Scientific™

Catalog Number p-4258997
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
19989-66-3
C8H7NOS
165.21
OZGXSRLIKDPNMX-UHFFFAOYSA-N
benzo d thiazol-6-ylmethanol, 6-hydroxymethyl benzothiazole, 6-benzothiazolemethanol, benzothiazol-6-yl-methanol, benzothiazol-6-yl methanol, benzothiazole-6-methanol, benzothiazol-6-ylmethanol, 6-hydroxymethylbenzothiazole, benzo d thiazol-6-yl-methanol
17860385
1,3-benzothiazol-6-ylmethanol
C1=CC2=C(C=C1CO)SC=N2
This item is not returnable. View return policy
19989-66-3
C8H7NOS
165.21
OZGXSRLIKDPNMX-UHFFFAOYSA-N
benzo d thiazol-6-ylmethanol, 6-hydroxymethyl benzothiazole, 6-benzothiazolemethanol, benzothiazol-6-yl-methanol, benzothiazol-6-yl methanol, benzothiazole-6-methanol, benzothiazol-6-ylmethanol, 6-hydroxymethylbenzothiazole, benzo d thiazol-6-yl-methanol
17860385
1,3-benzothiazol-6-ylmethanol
C1=CC2=C(C=C1CO)SC=N2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 103.0°C to 105.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Formula Weight 165.22
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 6-(Hydroxymethyl)benzothiazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'