Läs mer
6-Hydroxyquinoline-(1H)-2-one, TRC
Produktkod. 30397618
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30397618 250 mg -
30390896 1 g -
30394137 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30397618 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH953250250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19315-93-6
Molekylformel C9 H7 N O2
Molekylvikt (g/mol) 161.16
Synonym 2(1H)-Quinolinone, 6-hydroxy-, 2,6-Quinolinediol (7CI,8CI), 6-Hydroxy-2(1H)-quinolinone, 2,6-Dihydroxyquinoline, 6-Hydroxy-1H-quinolin-2-one, 6-Hydroxycarbostyril, 6-Hydroxyquinolin-2-one
IUPAC-namn quinoline-2,6-diol
LEDER Oc1ccc2nc(O)ccc2c1
Specifikationer
Kemiskt namn eller material 6-Hydroxyquinoline-(1H)-2-one
Kvantitet 250 mg
InChI formel InChI=1S/C9H7NO2/c11-7-2-3-8-6(5-7)1-4-9(12)10-8/h1-5,11H,(H,10,12)
Formel vikt 161.0477