Läs mer
6-Iodo Diosmin, TRC
Produktkod. 30009875
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009875 1 mg
30011752 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7058501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1431536-92-3
Molekylformel C28 H31 I O15
Molekylvikt (g/mol) 734.44
Synonym 7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6-iodo-4H-1-benzopyran-4-one, 6-Iododiosmin, Diosmin Imp. D (EP)
IUPAC-namn 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6-iodo-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
LEDER COc1ccc(cc1O)C2=CC(=O)c3c(O)c(I)c(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2
Specifikationer
Kemiskt namn eller material 6-Iodo Diosmin
Kvantitet 1 mg
InChI formel InChI=1S/C28H31IO15/c1-9-20(32)23(35)25(37)27(41-9)40-8-17-21(33)24(36)26(38)28(44-17)43-16-7-15-18(22(34)19(16)29)12(31)6-14(42-15)10-3-4-13(39-2)11(30)5-10/h3-7,9,17,20-21,23-28,30,32-38H,8H2,1-2H3/t9-,17+,20-,21+,23+,24-,25+,26+,27+,28+/m0/s1
Formel vikt 734.0708