Läs mer
6-Isopropyl-2-decahydronaphthalenol (Mixture of Diastereomers), TRC
Produktkod. 17904095
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17904095 50 mg
17950425 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17904095 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI86830550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34131-99-2
Molekylformel C13 H24 O
Molekylvikt (g/mol) 196.33
Synonym 2-Naphthol, decahydro-6-isopropyl- (8CI), Decahydro-6-(1-methylethyl)-2-naphthalenol, 6-Isopropyl-2-decahydronaphthalenol
IUPAC-namn 6-propan-2-yl-1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-ol
LEDER CC(C)C1CCC2CC(O)CCC2C1
Specifikationer
Kemiskt namn eller material 6-Isopropyl-2-decahydronaphthalenol
Kvantitet 50 mg
InChI formel InChI=1S/C13H24O/c1-9(2)10-3-4-12-8-13(14)6-5-11(12)7-10/h9-14H,3-8H2,1-2H3
Formel vikt 196.1827