Learn More
6-Isopropyl-4-oxo-4H-chromene-3-carbonitrile, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 50743-32-3 |
| Molecular Formula | C13H11NO2 |
| MDL Number | MFCD00191961 |
| Quantity | 25 g |
| SMILES | N#CC1=COC2=C(C=C(C(C)C)C=C2)C1=O |
| Molecular Weight (g/mol) | 213.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Isopropyl-4-oxo-4H-chromene-3-carbonitrile |