Läs mer
6-Keto Cholestanol, TRC
Produktkod. 17919184
Change view
Kvantitet:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17919184 1 g
17979554 250 mg
17997824 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17919184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK1850001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1175-06-0
Molekylformel C27 H46 O2
Molekylvikt (g/mol) 402.65
Synonym Cholestan-6-one, 3-hydroxy-, (3β,5α)-, 5α-Cholestan-6-one, 3β-hydroxy- (6CI,7CI,8CI), (3β,5α)-3-Hydroxycholestan-6-one, 3β-Hydroxy-5α-cholestan-6-one, 5α-Cholestan-3β-ol-6-one, 6-Ketocholestanol, 6-Oxocholestanol, NSC 178278, YS 164
IUPAC-namn (3S,5S,8S,9S,10R,13R,14S,17R)-3-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,4,5,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-6-one
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC(=O)[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 6-Ketocholestanol
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-16-25(29)24-15-19(28)11-13-27(24,5)23(20)12-14-26(21,22)4/h17-24,28H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,26-,27-/m1/s1
Formel vikt 402.3498
Procent renhet >95%