Läs mer
6-Keto Cyproterone Acetate, TRC
Produktkod. 30009446
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009446 25 mg
30007175 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009446 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK18765025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17184-05-3
Molekylformel C24 H30 O5
Molekylvikt (g/mol) 398.49
Synonym 3'H-Cyclopropa[1,2]pregna-1,4-diene-3,6,20-trione, 1β,2β-dihydro-17-hydroxy-, acetate (8CI), 1α,2α-Methylene-3,6,20-trioxopregna-4-en-17α-yl Acetate, 3,6,20-Trioxo-1β,2β-dihydro-3'H-cyclopropa[1,2]pregna-1,4-dien-17-yl Acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CC(=O)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 6-Keto Cyproterone Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C24H30O5/c1-12(25)24(29-13(2)26)8-6-16-14-10-21(28)19-11-20(27)15-9-18(15)23(19,4)17(14)5-7-22(16,24)3/h11,14-18H,5-10H2,1-4H3/t14-,15+,16-,17-,18-,22-,23-,24-/m0/s1
Formel vikt 398.2093
Procent renhet >95%