Läs mer
6-Ketoprogesterone, TRC
Produktkod. 30134961
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134961 500 mg
18010340 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134961 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK201005500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2243-08-5
Molekylformel C21 H28 O3
Molekylvikt (g/mol) 328.45
Synonym Pregn-4-ene-3,6,20-trione, 6-Ketoprogesterone, 6-Oxoprogesterone, NSC 76757, Progesterone 6-Oxo Impurity
IUPAC-namn (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,7,8,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthrene-3,6-dione
LEDER CC(=O)[C@H]1CC[C@H]2[C@@H]3CC(=O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 6-Ketoprogesterone
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C21H28O3/c1-12(22)15-4-5-16-14-11-19(24)18-10-13(23)6-8-21(18,3)17(14)7-9-20(15,16)2/h10,14-17H,4-9,11H2,1-3H3/t14-,15+,16-,17-,20+,21+/m0/s1
Formel vikt 328.2038
Procent renhet >95%