Läs mer
6-Maleimido-1-hexanol, TRC
Produktkod. 17984275
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17984275 250 mg
17940995 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17984275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM138560250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157503-18-9
Molekylformel C10 H15 N O3
Molekylvikt (g/mol) 197.23
Synonym 1-(6-Hydroxyhexyl)-1H-pyrrole-2,5-dione
IUPAC-namn 1-(6-hydroxyhexyl)pyrrole-2,5-dione
LEDER OCCCCCCN1C(=O)C=CC1=O
Specifikationer
Kemiskt namn eller material 6-Maleimido-1-hexanol
Kvantitet 250 mg
InChI formel InChI=1S/C10H15NO3/c12-8-4-2-1-3-7-11-9(13)5-6-10(11)14/h5-6,12H,1-4,7-8H2
Formel vikt 197.1052