Läs mer
6-Mercaptopurine Disulfide, TRC
Produktkod. 30134507
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134507 2.5 g
17920915 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134507 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2254552.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 49808-20-0
Molekylformel C10 H6 N8 S2
Molekylvikt (g/mol) 302.34
Synonym 6,6'-Dithiodi-9H-purine, 1H-Purine, 6,6'-dithiobis- (9CI), Purine, 6,6'-dithiodi- (7CI), 6,6'-Dithiodi-9H-purine, Bis(6-purinyl) disulfide, NSC 166569, NSC 77640, 6-Mercaptopurine Disulfide
IUPAC-namn 6-(7H-purin-6-yldisulfanyl)-9H-purine
LEDER S(Sc1ncnc2nc[nH]c12)c3ncnc4[nH]cnc34
Specifikationer
Kemiskt namn eller material 6-Mercaptopurine Disulfide
Kvantitet 2.5 g
InChI formel InChI=1S/C10H6N8S2/c1-11-5-7(13-1)15-3-17-9(5)19-20-10-6-8(14-2-12-6)16-4-18-10/h1-4H,(H,11,13,15,17)(H,12,14,16,18)
Formel vikt 302.0157