Learn More
6-Methoxy-2-methylbenzo[d]thiazole, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2941-72-2 |
| Molecular Formula | C9H9NOS |
| MDL Number | MFCD00005795 |
| Quantity | 1 g |
| SMILES | CC1=NC2=CC=C(OC)C=C2S1 |
| Molecular Weight (g/mol) | 179.24 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Methoxy-2-methylbenzo[d]thiazole |