Läs mer
6-Methoxy-2-naphthaleneacetic Acid (Desmethyl Naproxen), TRC
Produktkod. 18011850
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18011850 100 mg
18063550 10 mg
18010070 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18011850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM264695100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23981-47-7
Molekylformel C13 H12 O3
Molekylvikt (g/mol) 216.23
Synonym 2-Naphthaleneacetic acid, 6-methoxy-, 6-Methoxy-2-naphthaleneacetic acid, 2-(2-Methoxynaphthalen-6-yl)acetic acid, 2-(6-Methoxy-2-naphthyl)acetic acid, 6-Methoxy-2-naphthylacetic acid, BRL 10720, (6-Methoxy-2-naphthyl)acetic acid, (6-Methoxynaphthalen-2-yl)acetic acid, α-Demethylnaproxen, Naproxen Imp. I (EP)
IUPAC-namn 2-(6-methoxynaphthalen-2-yl)acetic acid
LEDER COc1ccc2cc(CC(=O)O)ccc2c1
Specifikationer
Kemiskt namn eller material alpha-Demethylnaproxen
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H12O3/c1-16-12-5-4-10-6-9(7-13(14)15)2-3-11(10)8-12/h2-6,8H,7H2,1H3,(H,14,15)
Formel vikt 216.0786
Procent renhet >95%