Läs mer
6-Methoxy-2-naphthol, TRC
Produktkod. 30466802
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466802 10 g
30476367 5 g
30472107 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30466802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26289010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5111-66-0
Molekylformel C11 H10 O2
Molekylvikt (g/mol) 174.2
Synonym 6-Methoxynaphthalen-2-ol, Naproxen Imp. H (EP), Hydroxynerolin
IUPAC-namn 6-methoxynaphthalen-2-ol
LEDER COc1ccc2cc(O)ccc2c1
Specifikationer
Kemiskt namn eller material 6-Methoxynaphthalen-2-ol (Hydroxynerolin)
Kvantitet 10 g
InChI formel InChI=1S/C11H10O2/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7,12H,1H3
Formel vikt 174.0681