Learn More
6-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2472-22-2 |
| Molecular Formula | C11H12O2 |
| MDL Number | MFCD00001729 |
| Quantity | 1 g |
| SMILES | O=C1CC2=C(C=C(OC)C=C2)CC1 |
| Molecular Weight (g/mol) | 176.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Methoxy-3,4-dihydronaphthalen-2(1H)-one |