Läs mer
6-Methoxy-m-cresol, TRC
Produktkod. 30476993
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30476993 250 mg -
30474934 2.5 g 2.5g
30474129 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476993 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM271880250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1195-09-1
Molekylformel C8H10O2
Molekylvikt (g/mol) 138.16
Synonym Isocreosol, 2-Hydroxy-4-methylanisole, 3-Hydroxy-4-methoxy-1-methylbenzene, 3-Hydroxy-4-methoxytoluene, 5-Methyl-2-methoxyphenol, 5-Methylguaiacol, o-Hydroxy-p-methylanisole
IUPAC-namn 2-methoxy-5-methylphenol
LEDER COc1ccc(C)cc1O
Specifikationer
Kemiskt namn eller material 6-Methoxy-m-cresol
Kvantitet 250 mg
InChI formel InChI=1S/C8H10O2/c1-6-3-4-8(10-2)7(9)5-6/h3-5,9H,1-2H3
Formel vikt 138.0681