Läs mer
6-Methyl-2,3'-bipyridine, TRC
Produktkod. 30482451
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482451 100 mg
30465513 250 mg
30464541 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30482451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM296433100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78210-78-3
Molekylformel C11H10N2
Molekylvikt (g/mol) 170.21
Synonym 6-Methyl-2,3'-bipyridine, 2-Methyl-6-(pyridin-3-yl)pyridine, 2-Methyl-6-pyridin-3-ylpyridine
LEDER Cc1cccc(n1)c2cccnc2
Specifikationer
Kemiskt namn eller material 6-Methyl-2,3'-bipyridine
Kvantitet 100 mg
InChI formel InChI=1S/C11H10N2/c1-9-4-2-6-11(13-9)10-5-3-7-12-8-10/h2-8H,1H3
Formel vikt 170.0844