Läs mer
6-Methyl-2-oxo-5-(4-pyridyl)-1H-pyridine-3-carboxamide, Mikromol™
Produktkod. 30052662
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052662 100 mg
30050784 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052662 Leverantör Mikromol Leverantörsnummer MM1539.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methyl-2-oxo-5-(4-pyridyl)-1H-pyridine-3-carboxamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Methyl-2-oxo-5-(4-pyridyl)-1H-pyridine-3-carboxamide
CAS 80047-24-1
Analyt- eller komponentnamn 6-Methyl-2-oxo-5-(4-pyridyl)-1H-pyridine-3-carboxamide
Frakt skick Room Temperature
Molekylformel C12 H11 N3 O2
Certifikat och efterlevnad ISO 17025
Synonym [3,4'-Bipyridine]-5-carboxamide, 1,6-dihydro-2-methyl-6-oxo-, Win 47338, Milrinone Related Compound A
InChI formel InChI=1S/C12H11N3O2/c1-7-9(8-2-4-14-5-3-8)6-10(11(13)16)12(17)15-7/h2-6H,1H3,(H2,13,16)(H,15,17)
LEDER CC1=C(C=C(C(=O)N)C(=O)N1)c2ccncc2
IUPAC-namn 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carboxamide
Molekylvikt (g/mol) 229.23
Kvantitet 100 mg
Formel vikt 229.0851 g/mol