Läs mer
6α-Methyl-20(R)-hydroxy Prednisolone 21-Acetate, TRC
Produktkod. 30466064
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30466064 10 mg
1 options
Produktkod. 30466064 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31295510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6alpha-Methyl-20(R)-hydroxy Prednisolone 21-Acetate
CAS 93963-74-7
Molekylformel C24H34O6
Kvantitet 10 mg
Synonym (6α,11β,20R)-21-(Acetyloxy)-11,17,20-trihydroxy-6-methylpregna-1,4-dien-3-one, Methylprednisolone Acetate EP Impurity A
InChI formel InChI=1S/C24H34O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19-21,27-29H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,20+,21+,22-,23-,24-/m0/s1
LEDER [H][C@@]12[C@]([C@](C=C3)(C)C([C@@H](C)C2)=CC3=O)([H])[C@@H](O)C[C@@]4(C)[C@@]1([H])CC[C@@]4([C@H](O)COC(C)=O)O
Molekylvikt (g/mol) 418.52
Formel vikt 418.2355