Läs mer
6-Methyl-4-phenylchroman-2-one, Mikromol™
Produktkod. 30457766
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30457766 4 x 25 mg
30051226 100 mg
30050386 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30457766 Leverantör Mikromol Leverantörsnummer MM0900.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methyl-4-phenylchroman-2-one, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Methyl-4-phenylchroman-2-one
CAS 40546-94-9
Analyt- eller komponentnamn 6-Methyl-4-phenylchroman-2-one
Frakt skick Room Temperature
Molekylformel C16 H14 O2
Certifikat och efterlevnad ISO 17025
Synonym 2H-1-Benzopyran-2-one, 3,4-dihydro-6-methyl-4-phenyl-, Hydrocoumarin, 6-methyl-4-phenyl- (6CI,7CI), 3,4-Dihydro-6-methyl-4-phenyl-2H-benzopyran-2-one, 6-Methyl-4-phenyl-3,4-dihydro-1-benzopyran-2-one, 6-Methyl-4-phenyl-3,4-dihydrocoumarin, 6-Methyl-4-phenylchroman-2-one
InChI formel InChI=1S/C16H14O2/c1-11-7-8-15-14(9-11)13(10-16(17)18-15)12-5-3-2-4-6-12/h2-9,13H,10H2,1H3
Lösningstyp Neat
LEDER Cc1ccc2OC(=O)CC(c3ccccc3)c2c1
IUPAC-namn 6-methyl-4-phenyl-3,4-dihydrochromen-2-one
Molekylvikt (g/mol) 238.28
Kvantitet 4 x 25 mg
Formel vikt 238.0994 g/mol