Läs mer
6-Methylmercaptopurine, TRC
Produktkod. 17951845
Change view
Kvantitet:
10 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17951845 250 mg
17901915 1 g
17910935 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17951845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM320270250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50-66-8
Molekylformel C6 H6 N4 S
Molekylvikt (g/mol) 166.2
Synonym 6-(Methylthio)-9H-purine, 6-(Methylthio)purine, NSC 20105, S-Methyl-6-mercaptopurine, SQ 8343;
IUPAC-namn 6-methylsulfanyl-7H-purine
LEDER CSc1ncnc2nc[nH]c12
Specifikationer
Kemiskt namn eller material 6-Methylmercaptopurine
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C6H6N4S/c1-11-6-4-5(8-2-7-4)9-3-10-6/h2-3H,1H3,(H,7,8,9,10)
Formel vikt 166.0313
Procent renhet >95%