Läs mer
6-Methyloctanoic Acid, TRC
Produktkod. 30468495
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468495 50 mg
30483227 100 mg
30469615 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30468495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM02134050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 504-99-4
Molekylformel C9H18O2
Molekylvikt (g/mol) 158.24
Synonym 6-methyl-Octanoic acid, (±)-6-Methyloctanoic acid, 6-Methylcaprylic acid, Anteisononanoic acid, ai-Nonanoic acid
IUPAC-namn 6-methyloctanoic acid
LEDER CCC(C)CCCCC(=O)O
Specifikationer
Kemiskt namn eller material 6-Methyloctanoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C9H18O2/c1-3-8(2)6-4-5-7-9(10)11/h8H,3-7H2,1-2H3,(H,10,11)
Formel vikt 158.1307