Läs mer
6-Methylpterin, TRC
Produktkod. 30132629
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132629 1 g
30004287 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132629 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4779051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 708-75-8
Molekylformel C7 H7 N5 O
Molekylvikt (g/mol) 177.16
Synonym 4(3H)-Pteridinone, 2-amino-6-methyl-, 4(1H)-Pteridinone, 2-amino-6-methyl- (8CI,9CI), 2-Amino-6-methyl-4(3H)-pteridinone, 2-Amino-4-hydroxy-6-methylpteridine, 6-Methylpterin
IUPAC-namn 2-amino-6-methyl-1H-pteridin-4-one
LEDER Cc1cnc2NC(=NC(=O)c2n1)N
Specifikationer
Kemiskt namn eller material 6-Methylpterin
Kvantitet 1 g
InChI formel InChI=1S/C7H7N5O/c1-3-2-9-5-4(10-3)6(13)12-7(8)11-5/h2H,1H3,(H3,8,9,11,12,13)
Formel vikt 177.0651