Läs mer
6-Methylpyrazinecarboxylic Acid, TRC
Produktkod. 30474815
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30474815 500 mg
1 options
Produktkod. 30474815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM327375500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Methylpyrazinecarboxylic Acid
CAS 5521-61-9
Molekylformel C6H6N2O2
Kvantitet 500 mg
Synonym 6-Methyl-2-pyrazinecarboxylic Acid, 2-Methylpyrazine-6-carboxylic Acid
InChI formel InChI=1S/C6H6N2O2/c1-4-2-7-3-5(8-4)6(9)10/h2-3H,1H3,(H,9,10)
LEDER Cc1cncc(n1)C(=O)O
Molekylvikt (g/mol) 138.12